Assay Details
Gene Symbol: ZNF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK123799.1 | - | 1 | 43 | 93 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | zinc finger and SCAN domain containing 21 | ZSCAN21 |
NY-REN-21, ZNF38, Zipro1 |
Anti-sense | Chr. 7 - 100049770 - 100065041 on NCBI Build GRCh38 | |
Downstream | COP9 signalosome subunit 6 | COPS6 |
CSN6, MOV34-34KD |
23.4kb | Anti-sense | Chr. 7 - 100088960 - 100092200 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
