Assay Details
| Gene Symbol: FAN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK026787.1 | - | 1 | 1450 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | hect domain and RLD 2 pseudogene 10 | HERC2P10 | -98.7kb | Sense | Chr. 15 - 30818036 - 30827783 on NCBI Build GRCh38 | |
| Downstream | transient receptor potential cation channel subfamily M member 1 | TRPM1 |
CSNB1C, LTRPC1, MLSN1 |
72.7kb | Anti-sense | Chr. 15 - 31001061 - 31161273 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

FAN1
Genomic Map