Assay Details
| Gene Symbol: RPE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006712677.3 | XP_006712740.1 | 7 | 5188 | 86 | |
| GenBank mRNA | AK055724.1 | - | 1 | 1842 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | KAT8 regulatory NSL complex subunit 1 like | KANSL1L |
C2orf67, MSL1v2 |
Anti-sense | Chr. 2 - 210021421 - 210171370 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101928020 | LOC101928020 | 5.4kb | Sense | Chr. 2 - 210029922 - 210064356 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

RPE
Genomic Map