Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 143 | TMEM143 | Sense | Chr. 19 - 48332356 - 48364237 on NCBI Build GRCh38 | ||
| Downstream | synaptogyrin 4 | SYNGR4 | 26.7kb | Anti-sense | Chr. 19 - 48364281 - 48376377 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map