Assay Details
| Gene Symbol: ALPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052947.3 | NP_443179.3 | 4 | 1150 | 93 | |
| XM_011525801.2 | XP_011524103.1 | 4 | 1383 | 93 | ||
| XM_011525802.2 | XP_011524104.1 | 2 | 1770 | 93 | ||
| XM_017025535.1 | XP_016881024.1 | 4 | 1384 | 93 | ||
| XM_017025536.1 | XP_016881025.1 | 4 | 1384 | 93 | ||
| XM_017025537.1 | XP_016881026.1 | 4 | 1385 | 93 | ||
| GenBank mRNA | AK125243.1 | - | 1 | 330 | 93 | |
| BX647796.1 | - | 4 | 1151 | 93 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000542.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR18005652.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005652.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005652.hg.1 | TC18000542.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005653.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005653.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005653.hg.1 | TC18000542.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005655.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005655.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005655.hg.1 | TC18000542.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005651.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005651.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005651.hg.1 | TC18000542.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000587399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000587399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000587399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000587399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000587842.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000587842.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000587842.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000587842.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000589204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000589204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000589204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000590642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000590642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000590662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000590662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000590662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | ENST00000590662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800160621.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800160621.hg.1 | ENST00000590642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800160621.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800160621.hg.1 | TC1800008804.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800160621.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800160621.hg.1 | ENST00000590642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800160621.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800160623.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800160623.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800160623.hg.1 | TC1800008804.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800160623.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800160623.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800160626.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800160626.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800160626.hg.1 | TC1800008804.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800160626.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800160626.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000587399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000587399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000587399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000587399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000587842.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000587842.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000587842.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000587842.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000589204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000589204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000589204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000590642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000590642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000590662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000590662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000590662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | ENST00000590662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008804.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800160619.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800160619.hg.1 | ENST00000590662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800160619.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800160619.hg.1 | TC1800008804.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800160619.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800160619.hg.1 | ENST00000590662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800160619.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800160620.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800160620.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800160620.hg.1 | TC1800008804.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800160620.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800160620.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800160624.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800160624.hg.1 | NM_052947.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800160624.hg.1 | TC1800008804.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800160624.hg.1 | ENST00000361673.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800160624.hg.1 | NM_052947.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 156577 | - | - |

ALPK2
Genomic Map