Assay Details
| Gene Symbol: LOC644285 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_130929.1 | 1 | 743 | 98 | ||
| GenBank mRNA | AK126853.1 | - | 1 | 744 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC644285
Genomic Map