| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18005413.hg.1 |
NM_016626.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18005413.hg.1 |
TC18000517.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18005411.hg.1 |
ENST00000406189.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18005411.hg.1 |
NM_016626.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18005411.hg.1 |
TC18000517.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008707.hg.1 |
ENST00000406189.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008707.hg.1 |
ENST00000406189.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008707.hg.1 |
ENST00000406189.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008707.hg.1 |
ENST00000406189.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008707.hg.1 |
ENST00000406189.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008707.hg.1 |
ENST00000406189.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008707.hg.1 |
ENST00000591040.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008707.hg.1 |
ENST00000591040.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008707.hg.1 |
ENST00000592416.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008707.hg.1 |
ENST00000592416.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008707.hg.1 |
ENST00000616921.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008707.hg.1 |
ENST00000616921.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008707.hg.1 |
ENST00000616921.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008707.hg.1 |
ENST00000616921.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008707.hg.1 |
ENST00000616921.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008707.hg.1 |
NM_016626.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008707.hg.1 |
NM_016626.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008707.hg.1 |
NM_016626.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008707.hg.1 |
NM_016626.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008707.hg.1 |
NM_016626.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008707.hg.1 |
NM_016626.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008707.hg.1 |
NM_016626.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18005412.hg.1 |
ENST00000406189.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18005412.hg.1 |
NM_016626.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18005412.hg.1 |
TC18000517.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18005410.hg.1 |
ENST00000406189.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18005410.hg.1 |
NM_016626.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18005410.hg.1 |
TC18000517.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1800159778.hg.1 |
ENST00000406189.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800159778.hg.1 |
ENST00000591040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800159778.hg.1 |
ENST00000616921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800159778.hg.1 |
NM_016626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800159778.hg.1 |
TC1800008707.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800159778.hg.1 |
ENST00000406189.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800159778.hg.1 |
ENST00000591040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800159778.hg.1 |
ENST00000616921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800159778.hg.1 |
NM_016626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800159785.hg.1 |
NM_016626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800159785.hg.1 |
TC1800008707.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800159785.hg.1 |
NM_016626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800159783.hg.1 |
ENST00000406189.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800159783.hg.1 |
NM_016626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800159783.hg.1 |
TC1800008707.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800159783.hg.1 |
ENST00000406189.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800159783.hg.1 |
NM_016626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008707.hg.1 |
ENST00000406189.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008707.hg.1 |
ENST00000406189.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008707.hg.1 |
ENST00000406189.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008707.hg.1 |
ENST00000406189.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008707.hg.1 |
ENST00000406189.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008707.hg.1 |
ENST00000406189.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008707.hg.1 |
ENST00000591040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008707.hg.1 |
ENST00000591040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008707.hg.1 |
ENST00000592416.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008707.hg.1 |
ENST00000592416.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008707.hg.1 |
ENST00000616921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008707.hg.1 |
ENST00000616921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008707.hg.1 |
ENST00000616921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008707.hg.1 |
ENST00000616921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008707.hg.1 |
ENST00000616921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008707.hg.1 |
NM_016626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008707.hg.1 |
NM_016626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008707.hg.1 |
NM_016626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008707.hg.1 |
NM_016626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008707.hg.1 |
NM_016626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008707.hg.1 |
NM_016626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008707.hg.1 |
NM_016626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800159784.hg.1 |
ENST00000406189.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800159784.hg.1 |
ENST00000616921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800159784.hg.1 |
NM_016626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800159784.hg.1 |
TC1800008707.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800159784.hg.1 |
ENST00000406189.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800159784.hg.1 |
ENST00000616921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800159784.hg.1 |
NM_016626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800159780.hg.1 |
ENST00000406189.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800159780.hg.1 |
ENST00000592416.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800159780.hg.1 |
ENST00000616921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800159780.hg.1 |
NM_016626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800159780.hg.1 |
TC1800008707.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800159780.hg.1 |
ENST00000406189.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800159780.hg.1 |
ENST00000592416.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800159780.hg.1 |
ENST00000616921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800159780.hg.1 |
NM_016626.4 |
Refseq |