| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005681.hg.1 |
ENST00000171757.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005681.hg.1 |
ENST00000475374.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005681.hg.1 |
ENST00000544091.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005681.hg.1 |
NM_014499.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005681.hg.1 |
NM_198333.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005681.hg.1 |
TC0X000427.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
ENST00000171757.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
ENST00000171757.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
ENST00000171757.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
ENST00000171757.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
ENST00000171757.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
ENST00000171757.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
ENST00000171757.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
ENST00000461541.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
ENST00000461541.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
ENST00000475374.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
ENST00000475374.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
ENST00000475374.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
ENST00000475374.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
ENST00000544091.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
ENST00000544091.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
ENST00000544091.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
ENST00000544091.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
ENST00000544091.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
ENST00000544091.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
NM_014499.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
NM_014499.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
NM_014499.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
NM_014499.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
NM_014499.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
NM_014499.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
NM_014499.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
NM_014499.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
NM_198333.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
NM_198333.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
NM_198333.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
NM_198333.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
NM_198333.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007716.hg.1 |
NM_198333.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00152868.hg.1 |
ENST00000171757.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00152868.hg.1 |
ENST00000475374.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00152868.hg.1 |
ENST00000544091.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00152868.hg.1 |
NM_014499.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00152868.hg.1 |
NM_198333.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00152868.hg.1 |
ENST00000171757.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00152868.hg.1 |
ENST00000475374.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00152868.hg.1 |
ENST00000544091.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00152868.hg.1 |
NM_014499.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00152868.hg.1 |
NM_198333.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00152868.hg.1 |
TC0X00007716.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
ENST00000171757.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
ENST00000171757.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
ENST00000171757.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
ENST00000171757.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
ENST00000171757.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
ENST00000171757.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
ENST00000171757.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
ENST00000461541.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
ENST00000461541.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
ENST00000475374.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
ENST00000475374.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
ENST00000475374.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
ENST00000475374.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
ENST00000544091.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
ENST00000544091.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
ENST00000544091.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
ENST00000544091.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
ENST00000544091.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
ENST00000544091.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
NM_014499.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
NM_014499.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
NM_014499.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
NM_014499.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
NM_014499.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
NM_014499.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
NM_014499.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
NM_014499.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
NM_198333.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
NM_198333.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
NM_198333.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
NM_198333.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
NM_198333.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007716.hg.1 |
NM_198333.2 |
Refseq |