Assay Details
| Gene Symbol: ZNF347 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172674.1 | NP_001166145.1 | 4 - 5 | 702 | 95 | |
| NM_001172675.1 | NP_001166146.1 | 4 - 5 | 382 | 95 | ||
| NM_032584.2 | NP_115973.2 | 4 - 5 | 379 | 95 | ||
| XM_005259335.4 | XP_005259392.1 | 4 - 5 | 701 | 95 | ||
| XM_005259336.4 | XP_005259393.1 | 4 - 5 | 491 | 95 | ||
| XM_017027384.1 | XP_016882873.1 | 4 - 5 | 721 | 95 | ||
| XM_017027385.1 | XP_016882874.1 | 4 - 5 | 405 | 95 | ||
| XM_017027386.1 | XP_016882875.1 | 3 - 4 | 327 | 95 | ||
| GenBank mRNA | AK096623.1 | - | 4 - 5 | 704 | 95 | |
| AK289994.1 | - | 4 - 5 | 366 | 95 | ||
| AK314603.1 | - | 4 - 5 | 363 | 95 | ||
| AY029765.1 | - | 4 - 5 | 341 | 95 | ||
| BC136255.1 | - | 3 - 4 | 331 | 9020242 | 95 | |
| BC143412.1 | - | 3 - 4 | 334 | 95 | ||
More Information
Set Membership: |
|

ZNF347
Genomic Map