| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05026981.hg.1 |
ENST00000274599.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05026981.hg.1 |
ENST00000394226.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05026981.hg.1 |
ENST00000418587.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05026981.hg.1 |
ENST00000427179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05026981.hg.1 |
ENST00000446148.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05026981.hg.1 |
NM_001172831.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05026981.hg.1 |
NM_001172832.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05026981.hg.1 |
NM_052860.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05026981.hg.1 |
TC05001944.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0500187064.hg.1 |
ENST00000274599.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187064.hg.1 |
ENST00000394226.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187064.hg.1 |
ENST00000418587.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187064.hg.1 |
ENST00000427179.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187064.hg.1 |
ENST00000446148.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187064.hg.1 |
NM_001172831.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500187064.hg.1 |
NM_001172832.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500187064.hg.1 |
NM_052860.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500187064.hg.1 |
ENST00000274599.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187064.hg.1 |
ENST00000394226.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187064.hg.1 |
ENST00000418587.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187064.hg.1 |
ENST00000427179.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187064.hg.1 |
ENST00000446148.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500187064.hg.1 |
NM_001172831.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500187064.hg.1 |
NM_001172832.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500187064.hg.1 |
NM_052860.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500187064.hg.1 |
TC0500012491.hg.1 |
Clariom D TC |