| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04018365.hg.1 |
ENST00000295470.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04018365.hg.1 |
ENST00000502762.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04018365.hg.1 |
ENST00000514511.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04018365.hg.1 |
NM_031372.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04018365.hg.1 |
NR_003249.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04018365.hg.1 |
TC04001334.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017148.hg.1 |
ENST00000427435.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176100.hg.1 |
ENST00000295470.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176100.hg.1 |
ENST00000349655.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176100.hg.1 |
ENST00000502762.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176100.hg.1 |
ENST00000507721.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176100.hg.1 |
ENST00000514511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176100.hg.1 |
ENST00000602300.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176100.hg.1 |
ENST00000621267.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176100.hg.1 |
ENST00000630114.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176100.hg.1 |
ENST00000630827.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176100.hg.1 |
NM_031372.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400176100.hg.1 |
NR_003249.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400176100.hg.1 |
ENST00000295470.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176100.hg.1 |
ENST00000349655.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176100.hg.1 |
ENST00000502762.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176100.hg.1 |
ENST00000507721.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176100.hg.1 |
ENST00000514511.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176100.hg.1 |
ENST00000602300.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176100.hg.1 |
ENST00000621267.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176100.hg.1 |
ENST00000630114.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176100.hg.1 |
ENST00000630827.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176100.hg.1 |
NM_031372.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400176100.hg.1 |
NR_003249.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400176100.hg.1 |
TC0400011173.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100017148.hg.1 |
ENST00000427435.1 |
Ensembl |