| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009901.hg.1 |
ENST00000321961.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009901.hg.1 |
ENST00000623849.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009901.hg.1 |
ENST00000624469.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009901.hg.1 |
ENST00000624469.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009901.hg.1 |
ENST00000624469.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009901.hg.1 |
NM_001005237.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016707.hg.1 |
ENST00000321961.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016707.hg.1 |
NM_001005237.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11016707.hg.1 |
TC11001319.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1100174801.hg.1 |
ENST00000321961.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174801.hg.1 |
ENST00000623849.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174801.hg.1 |
ENST00000624469.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174801.hg.1 |
NM_001005237.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100174801.hg.1 |
ENST00000321961.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174801.hg.1 |
ENST00000623849.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174801.hg.1 |
ENST00000624469.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174801.hg.1 |
NM_001005237.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100174801.hg.1 |
TC1100009901.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100009901.hg.1 |
ENST00000321961.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009901.hg.1 |
ENST00000623849.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009901.hg.1 |
ENST00000624469.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009901.hg.1 |
ENST00000624469.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009901.hg.1 |
ENST00000624469.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009901.hg.1 |
NM_001005237.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100174802.hg.1 |
ENST00000624469.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174802.hg.1 |
ENST00000624469.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100174802.hg.1 |
TC1100009901.hg.1 |
Clariom D TC |