Assay Details
Gene Symbol: LIG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098268.1 | NP_001091738.1 | 1 - 2 | 207 | 112 | |
GenBank mRNA | BX648833.1 | - | 1 - 2 | 208 | 112 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC13000867.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001098268.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
single strand break repair
in utero embryonic development
pro-B cell differentiation
DNA ligation
lagging strand elongation
nucleotide-excision repair, DNA gap filling
double-strand break repair
double-strand break repair via nonhomologous end joining
cell cycle
central nervous system development
cell proliferation
response to X-ray
response to gamma radiation
T cell differentiation in thymus
V(D)J recombination
immunoglobulin V(D)J recombination
T cell receptor V(D)J recombination
somatic stem cell population maintenance
negative regulation of neuron apoptotic process
isotype switching
positive regulation of fibroblast proliferation
positive regulation of neurogenesis
DNA ligation involved in DNA recombination
DNA ligation involved in DNA repair
chromosome organization
cell division
neuron apoptotic process
cellular response to lithium ion
DNA biosynthetic process
establishment of integrated proviral latency
double-strand break repair via classical nonhomologous end joining
positive regulation of chromosome organization
DNA binding
DNA ligase activity
DNA ligase (ATP) activity
protein binding
ATP binding
protein C-terminus binding
ligase activity
metal ion binding
single strand break repair
in utero embryonic development
pro-B cell differentiation
DNA ligation
lagging strand elongation
nucleotide-excision repair, DNA gap filling
double-strand break repair
double-strand break repair via nonhomologous end joining
cell cycle
central nervous system development
cell proliferation
response to X-ray
response to gamma radiation
T cell differentiation in thymus
V(D)J recombination
immunoglobulin V(D)J recombination
T cell receptor V(D)J recombination
somatic stem cell population maintenance
negative regulation of neuron apoptotic process
isotype switching
positive regulation of fibroblast proliferation
positive regulation of neurogenesis
DNA ligation involved in DNA recombination
DNA ligation involved in DNA repair
chromosome organization
cell division
neuron apoptotic process
cellular response to lithium ion
DNA biosynthetic process
establishment of integrated proviral latency
double-strand break repair via classical nonhomologous end joining
positive regulation of chromosome organization
DNA binding
DNA ligase activity
DNA ligase (ATP) activity
protein binding
ATP binding
protein C-terminus binding
ligase activity
metal ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | ENST00000356922.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | ENST00000356922.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | ENST00000356922.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | ENST00000356922.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | ENST00000356922.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | ENST00000356922.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | ENST00000405925.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | ENST00000405925.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | ENST00000405925.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | ENST00000405925.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | ENST00000405925.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | ENST00000442234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | ENST00000442234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | ENST00000442234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | ENST00000442234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | ENST00000442234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | ENST00000442234.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | ENST00000611712.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | ENST00000611712.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | ENST00000611712.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | ENST00000611712.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | ENST00000611712.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | ENST00000611712.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | ENST00000611712.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | ENST00000611712.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | ENST00000614526.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | ENST00000614526.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | ENST00000614526.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | ENST00000614526.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | NM_001098268.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | NM_001098268.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | NM_001098268.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | NM_001098268.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | NM_001098268.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | NM_002312.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | NM_002312.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | NM_002312.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | NM_002312.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | NM_002312.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | NM_002312.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | NM_206937.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | NM_206937.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | NM_206937.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | NM_206937.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | NM_206937.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009740.hg.1 | NM_206937.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13009691.hg.1 | ENST00000405925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13009691.hg.1 | NM_001098268.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13009691.hg.1 | TC13000867.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | ENST00000356922.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | ENST00000356922.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | ENST00000356922.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | ENST00000356922.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | ENST00000356922.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | ENST00000356922.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | ENST00000405925.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | ENST00000405925.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | ENST00000405925.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | ENST00000405925.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | ENST00000405925.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | ENST00000442234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | ENST00000442234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | ENST00000442234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | ENST00000442234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | ENST00000442234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | ENST00000442234.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | ENST00000611712.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | ENST00000611712.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | ENST00000611712.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | ENST00000611712.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | ENST00000611712.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | ENST00000611712.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | ENST00000611712.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | ENST00000611712.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | ENST00000614526.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | ENST00000614526.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | ENST00000614526.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | ENST00000614526.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | NM_001098268.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | NM_001098268.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | NM_001098268.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | NM_001098268.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | NM_001098268.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | NM_002312.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | NM_002312.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | NM_002312.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | NM_002312.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | NM_002312.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | NM_002312.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | NM_206937.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | NM_206937.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | NM_206937.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | NM_206937.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | NM_206937.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300009740.hg.1 | NM_206937.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300165001.hg.1 | ENST00000405925.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300165001.hg.1 | NM_001098268.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300165001.hg.1 | ENST00000405925.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300165001.hg.1 | NM_001098268.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300165001.hg.1 | TC1300009740.hg.1 | Clariom D TC |
