Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14001210.hg.1 |
ENST00000408907.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14001210.hg.1 |
NM_001099780.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14001210.hg.1 |
TC14000140.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006672.hg.1 |
ENST00000408907.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006672.hg.1 |
ENST00000408907.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006672.hg.1 |
ENST00000408907.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006672.hg.1 |
ENST00000408907.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006672.hg.1 |
NM_001099780.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006672.hg.1 |
NM_001099780.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400006672.hg.1 |
NM_001099780.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400145627.hg.1 |
ENST00000408907.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400145627.hg.1 |
NM_001099780.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400145627.hg.1 |
ENST00000408907.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400145627.hg.1 |
NM_001099780.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400145627.hg.1 |
TC1400006672.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400006672.hg.1 |
ENST00000408907.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006672.hg.1 |
ENST00000408907.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006672.hg.1 |
ENST00000408907.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006672.hg.1 |
ENST00000408907.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400006672.hg.1 |
NM_001099780.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006672.hg.1 |
NM_001099780.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400006672.hg.1 |
NM_001099780.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400145626.hg.1 |
ENST00000408907.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400145626.hg.1 |
NM_001099780.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400145626.hg.1 |
ENST00000408907.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400145626.hg.1 |
NM_001099780.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400145626.hg.1 |
TC1400006672.hg.1 |
Clariom D TC |