| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008992.hg.1 |
ENST00000321545.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008992.hg.1 |
ENST00000321545.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008992.hg.1 |
ENST00000321545.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008992.hg.1 |
ENST00000321545.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008992.hg.1 |
ENST00000321545.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008992.hg.1 |
ENST00000321545.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008992.hg.1 |
ENST00000321545.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008992.hg.1 |
ENST00000321545.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008992.hg.1 |
NM_213598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008992.hg.1 |
NM_213598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008992.hg.1 |
NM_213598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008992.hg.1 |
NM_213598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008992.hg.1 |
NM_213598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008992.hg.1 |
NM_213598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008992.hg.1 |
NM_213598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008992.hg.1 |
NM_213598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19012835.hg.1 |
ENST00000321545.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19012835.hg.1 |
NM_213598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19012835.hg.1 |
TC19000938.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1900170750.hg.1 |
ENST00000321545.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900170750.hg.1 |
NM_213598.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900170750.hg.1 |
TC1900008992.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900170750.hg.1 |
ENST00000321545.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900170750.hg.1 |
NM_213598.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008992.hg.1 |
ENST00000321545.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008992.hg.1 |
ENST00000321545.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008992.hg.1 |
ENST00000321545.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008992.hg.1 |
ENST00000321545.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008992.hg.1 |
ENST00000321545.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008992.hg.1 |
ENST00000321545.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008992.hg.1 |
ENST00000321545.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008992.hg.1 |
ENST00000321545.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008992.hg.1 |
NM_213598.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008992.hg.1 |
NM_213598.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008992.hg.1 |
NM_213598.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008992.hg.1 |
NM_213598.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008992.hg.1 |
NM_213598.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008992.hg.1 |
NM_213598.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008992.hg.1 |
NM_213598.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008992.hg.1 |
NM_213598.3 |
Refseq |