| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011381.hg.1 |
ENST00000319296.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011381.hg.1 |
ENST00000319296.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011381.hg.1 |
ENST00000319296.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011381.hg.1 |
ENST00000319296.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011381.hg.1 |
ENST00000319296.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011381.hg.1 |
ENST00000319296.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011381.hg.1 |
ENST00000319296.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011381.hg.1 |
ENST00000434417.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011381.hg.1 |
ENST00000434417.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011381.hg.1 |
ENST00000434417.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011381.hg.1 |
ENST00000434417.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011381.hg.1 |
ENST00000434417.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011381.hg.1 |
ENST00000564550.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011381.hg.1 |
ENST00000564550.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011381.hg.1 |
NM_001142305.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011381.hg.1 |
NM_001142305.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011381.hg.1 |
NM_001142305.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011381.hg.1 |
NM_001142305.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011381.hg.1 |
NM_001142305.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011381.hg.1 |
NM_016643.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011381.hg.1 |
NM_016643.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011381.hg.1 |
NM_016643.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011381.hg.1 |
NM_016643.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011381.hg.1 |
NM_016643.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16005002.hg.1 |
ENST00000319296.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16005002.hg.1 |
ENST00000434417.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16005002.hg.1 |
NM_001142305.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16005002.hg.1 |
NM_016643.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16005002.hg.1 |
TC16000347.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1600011381.hg.1 |
ENST00000319296.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011381.hg.1 |
ENST00000319296.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011381.hg.1 |
ENST00000319296.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011381.hg.1 |
ENST00000319296.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011381.hg.1 |
ENST00000319296.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011381.hg.1 |
ENST00000319296.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011381.hg.1 |
ENST00000319296.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011381.hg.1 |
ENST00000434417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011381.hg.1 |
ENST00000434417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011381.hg.1 |
ENST00000434417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011381.hg.1 |
ENST00000434417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011381.hg.1 |
ENST00000434417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011381.hg.1 |
ENST00000564550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011381.hg.1 |
ENST00000564550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011381.hg.1 |
NM_001142305.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011381.hg.1 |
NM_001142305.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011381.hg.1 |
NM_001142305.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011381.hg.1 |
NM_001142305.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011381.hg.1 |
NM_001142305.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011381.hg.1 |
NM_016643.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011381.hg.1 |
NM_016643.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011381.hg.1 |
NM_016643.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011381.hg.1 |
NM_016643.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011381.hg.1 |
NM_016643.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600190277.hg.1 |
ENST00000319296.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600190277.hg.1 |
ENST00000434417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600190277.hg.1 |
ENST00000564550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600190277.hg.1 |
NM_001142305.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600190277.hg.1 |
NM_016643.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600190277.hg.1 |
TC1600011381.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600190277.hg.1 |
ENST00000319296.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600190277.hg.1 |
ENST00000434417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600190277.hg.1 |
ENST00000564550.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600190277.hg.1 |
NM_001142305.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600190277.hg.1 |
NM_016643.3 |
Refseq |