Assay Details
| Gene Symbol: ZNF444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253792.1 | NP_001240721.1 | 3 - 4 | 685 | 88 | |
| NM_018337.3 | NP_060807.2 | 3 - 4 | 685 | 88 | ||
| XM_005259034.2 | XP_005259091.1 | 4 - 5 | 790 | 88 | ||
| XM_005259035.1 | XP_005259092.1 | 3 - 4 | 722 | 88 | ||
| XM_005259036.1 | XP_005259093.1 | 3 - 4 | 589 | 88 | ||
| XM_011527068.1 | XP_011525370.1 | 3 - 4 | 748 | 88 | ||
| GenBank mRNA | AB052954.1 | - | 3 - 4 | 528 | 88 | |
| AB052955.1 | - | 3 - 4 | 528 | 88 | ||
| AK001999.1 | - | 3 - 4 | 530 | 88 | ||
| AK223408.1 | - | 3 - 4 | 555 | 88 | ||
| AL834130.1 | - | 3 - 4 | 665 | 88 | ||
| BC021282.2 | - | 3 - 4 | 515 | 5016494 | 88 | |
More Information
Set Membership: |
|

ZNF444
Genomic Map