| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01035329.hg.1 |
ENST00000375121.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01035329.hg.1 |
NM_019062.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01035329.hg.1 |
TC01002305.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013159.hg.1 |
ENST00000375121.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013159.hg.1 |
ENST00000375121.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013159.hg.1 |
ENST00000375121.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013159.hg.1 |
ENST00000375121.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013159.hg.1 |
NM_019062.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013159.hg.1 |
NM_019062.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013159.hg.1 |
NM_019062.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013159.hg.1 |
NM_019062.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01035328.hg.1 |
ENST00000375121.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01035328.hg.1 |
NM_019062.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01035328.hg.1 |
TC01002305.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100197371.hg.1 |
ENST00000375121.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100197371.hg.1 |
NM_019062.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100197371.hg.1 |
ENST00000375121.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100197371.hg.1 |
NM_019062.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100197371.hg.1 |
TC0100013159.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100013159.hg.1 |
ENST00000375121.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013159.hg.1 |
ENST00000375121.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013159.hg.1 |
ENST00000375121.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013159.hg.1 |
ENST00000375121.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013159.hg.1 |
NM_019062.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013159.hg.1 |
NM_019062.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013159.hg.1 |
NM_019062.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013159.hg.1 |
NM_019062.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100197370.hg.1 |
ENST00000375121.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100197370.hg.1 |
NM_019062.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100197370.hg.1 |
ENST00000375121.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100197370.hg.1 |
NM_019062.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100197370.hg.1 |
TC0100013159.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100197369.hg.1 |
ENST00000375121.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100197369.hg.1 |
NM_019062.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100197369.hg.1 |
ENST00000375121.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100197369.hg.1 |
NM_019062.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100197369.hg.1 |
TC0100013159.hg.1 |
Clariom D TC |