| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008750.hg.1 |
ENST00000370278.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008750.hg.1 |
ENST00000370278.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008750.hg.1 |
ENST00000370278.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008750.hg.1 |
ENST00000370278.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008750.hg.1 |
ENST00000370278.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008750.hg.1 |
ENST00000370278.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008750.hg.1 |
ENST00000417212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008750.hg.1 |
ENST00000417212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008750.hg.1 |
ENST00000417212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008750.hg.1 |
ENST00000417212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008750.hg.1 |
ENST00000417212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008750.hg.1 |
ENST00000598245.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008750.hg.1 |
ENST00000598245.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008750.hg.1 |
ENST00000598245.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008750.hg.1 |
ENST00000598245.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008750.hg.1 |
ENST00000598245.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008750.hg.1 |
ENST00000598245.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008750.hg.1 |
ENST00000598245.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008750.hg.1 |
NM_005362.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008750.hg.1 |
NM_005362.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008750.hg.1 |
NM_005362.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008750.hg.1 |
NM_005362.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008750.hg.1 |
NM_005362.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008750.hg.1 |
NM_005362.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009530.hg.1 |
ENST00000329342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009530.hg.1 |
NM_005363.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009530.hg.1 |
NM_175868.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009530.hg.1 |
TC0X000739.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009535.hg.1 |
ENST00000329342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009535.hg.1 |
NM_005363.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009535.hg.1 |
NM_175868.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009535.hg.1 |
TC0X000739.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009531.hg.1 |
ENST00000329342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009531.hg.1 |
NM_005363.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009531.hg.1 |
NM_175868.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009531.hg.1 |
TC0X000739.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009532.hg.1 |
ENST00000329342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009532.hg.1 |
NM_005363.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009532.hg.1 |
NM_175868.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009532.hg.1 |
TC0X000739.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009534.hg.1 |
ENST00000329342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009534.hg.1 |
NM_005363.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009534.hg.1 |
NM_175868.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009534.hg.1 |
TC0X000739.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009533.hg.1 |
ENST00000329342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009533.hg.1 |
NM_005363.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009533.hg.1 |
NM_175868.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009533.hg.1 |
TC0X000739.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X019549.hg.1 |
ENST00000370278.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X019549.hg.1 |
ENST00000393902.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X019549.hg.1 |
NM_005362.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X019549.hg.1 |
TC0X001505.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0X00175489.hg.1 |
ENST00000329342.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175489.hg.1 |
ENST00000412733.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175489.hg.1 |
ENST00000457643.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175489.hg.1 |
ENST00000616035.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175489.hg.1 |
NM_005363.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175489.hg.1 |
NM_175868.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175489.hg.1 |
ENST00000329342.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175489.hg.1 |
ENST00000412733.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175489.hg.1 |
ENST00000457643.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175489.hg.1 |
ENST00000616035.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175489.hg.1 |
NM_005363.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175489.hg.1 |
NM_175868.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175489.hg.1 |
TC0X00011136.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00175486.hg.1 |
ENST00000329342.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175486.hg.1 |
ENST00000616035.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175486.hg.1 |
NM_005363.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175486.hg.1 |
NM_175868.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175486.hg.1 |
ENST00000329342.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175486.hg.1 |
ENST00000616035.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175486.hg.1 |
NM_005363.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175486.hg.1 |
NM_175868.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175486.hg.1 |
TC0X00011136.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00175487.hg.1 |
ENST00000329342.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175487.hg.1 |
ENST00000457643.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175487.hg.1 |
ENST00000616035.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175487.hg.1 |
NM_005363.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175487.hg.1 |
NM_175868.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175487.hg.1 |
ENST00000329342.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175487.hg.1 |
ENST00000457643.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175487.hg.1 |
ENST00000616035.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175487.hg.1 |
NM_005363.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175487.hg.1 |
NM_175868.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175487.hg.1 |
TC0X00011136.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00008750.hg.1 |
ENST00000370278.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008750.hg.1 |
ENST00000370278.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008750.hg.1 |
ENST00000370278.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008750.hg.1 |
ENST00000370278.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008750.hg.1 |
ENST00000370278.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008750.hg.1 |
ENST00000370278.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008750.hg.1 |
ENST00000417212.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008750.hg.1 |
ENST00000417212.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008750.hg.1 |
ENST00000417212.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008750.hg.1 |
ENST00000417212.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008750.hg.1 |
ENST00000417212.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008750.hg.1 |
ENST00000598245.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008750.hg.1 |
ENST00000598245.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008750.hg.1 |
ENST00000598245.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008750.hg.1 |
ENST00000598245.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008750.hg.1 |
ENST00000598245.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008750.hg.1 |
ENST00000598245.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008750.hg.1 |
ENST00000598245.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008750.hg.1 |
NM_005362.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008750.hg.1 |
NM_005362.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008750.hg.1 |
NM_005362.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008750.hg.1 |
NM_005362.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008750.hg.1 |
NM_005362.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008750.hg.1 |
NM_005362.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175484.hg.1 |
ENST00000329342.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175484.hg.1 |
ENST00000616035.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175484.hg.1 |
NM_005363.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175484.hg.1 |
NM_175868.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175484.hg.1 |
ENST00000329342.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175484.hg.1 |
ENST00000616035.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175484.hg.1 |
NM_005363.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175484.hg.1 |
NM_175868.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175484.hg.1 |
TC0X00011136.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00175485.hg.1 |
ENST00000329342.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175485.hg.1 |
ENST00000616035.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175485.hg.1 |
NM_005363.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175485.hg.1 |
NM_175868.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175485.hg.1 |
ENST00000329342.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175485.hg.1 |
ENST00000616035.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175485.hg.1 |
NM_005363.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175485.hg.1 |
NM_175868.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175485.hg.1 |
TC0X00011136.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00175488.hg.1 |
ENST00000329342.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175488.hg.1 |
ENST00000412733.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175488.hg.1 |
ENST00000457643.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175488.hg.1 |
ENST00000616035.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175488.hg.1 |
NM_005363.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175488.hg.1 |
NM_175868.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175488.hg.1 |
ENST00000329342.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175488.hg.1 |
ENST00000412733.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175488.hg.1 |
ENST00000457643.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175488.hg.1 |
ENST00000616035.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00175488.hg.1 |
NM_005363.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175488.hg.1 |
NM_175868.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00175488.hg.1 |
TC0X00011136.hg.1 |
Clariom D TC |