Assay Details
| Gene Symbol: MIR182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029614.1 | 1 | 54 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nuclear respiratory factor 1 | NRF1 | ALPHA-PAL | -13.3kb | Anti-sense | Chr. 7 - 129611702 - 129757082 on NCBI Build GRCh38 |
| Downstream | microRNA 96 | MIR96 |
DFNA50, MIRN96, hsa-mir-96, miR-96, miRNA96 |
4.2kb | Sense | Chr. 7 - 129774692 - 129774769 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0700012578.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07001854.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778724 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778717 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778746 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16017706 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26036571 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778740 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778725 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16017702 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26036569 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778737 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778736 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778731 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26036576 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778744 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778735 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778719 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16017698 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16017707 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778739 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778721 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778714 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26036573 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16017700 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778747 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778752 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778749 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16017708 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778713 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778733 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778741 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26036595 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778728 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778742 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778745 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778747 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778721 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778735 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778746 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778725 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778713 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26036569 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26036571 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16017700 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778724 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16017702 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778733 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778741 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26036573 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16017706 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778736 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26036576 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778742 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778745 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778737 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16017708 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778728 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778752 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778739 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16017698 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26036595 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778717 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16017707 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778731 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778719 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778744 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778714 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778740 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05778749 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.1 ST Array | 8142880 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR07028734.hg.1 | ENST00000385255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07028734.hg.1 | NR_029614.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07028734.hg.1 | TC07001854.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012578.hg.1 | ENST00000385255.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012578.hg.1 | NR_029614.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700190596.hg.1 | ENST00000385255.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700190596.hg.1 | NR_029614.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700190596.hg.1 | ENST00000385255.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700190596.hg.1 | NR_029614.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700190596.hg.1 | TC0700012578.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700012578.hg.1 | ENST00000385255.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012578.hg.1 | NR_029614.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A033531 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A033531 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3309560 | NR_029614.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3309560 | NR_029614.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3309560 | NR_029614.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3309560 | NR_029614.1 | human_transcriptome |

MIR182
Genomic Map