Assay Details
| Gene Symbol: MIR210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029623.1 | 1 | 55 | 61 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MIR210 host gene | MIR210HG | Sense | Chr. 11 - 565657 - 568457 on NCBI Build GRCh38 | ||
| Downstream | uncharacterized LOC143666 | LOC143666 | 5.6kb | Sense | Chr. 11 - 573808 - 575885 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595238 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27989534 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27989515 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27989541 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595256 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595258 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595255 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27989518 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595251 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595248 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595250 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595240 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595265 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595253 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595247 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595245 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595264 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595257 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595252 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16406391 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27989546 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595256 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27989515 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16406391 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595255 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595238 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595250 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595240 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595253 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595252 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27989518 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595251 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27989546 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595248 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595265 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595247 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27989534 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595264 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27989541 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595245 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595258 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595257 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.1 ST Array | 7945460 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009680.hg.1 | ENST00000362168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009680.hg.1 | ENST00000500447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009680.hg.1 | ENST00000500447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009680.hg.1 | ENST00000500447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009680.hg.1 | ENST00000500447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009680.hg.1 | ENST00000500447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009680.hg.1 | ENST00000528245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009680.hg.1 | ENST00000528245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009680.hg.1 | ENST00000528245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009680.hg.1 | ENST00000528245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009680.hg.1 | ENST00000528245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009680.hg.1 | ENST00000533920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009680.hg.1 | ENST00000533920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009680.hg.1 | ENST00000533920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009680.hg.1 | ENST00000533920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009680.hg.1 | ENST00000533920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009680.hg.1 | ENST00000534540.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009680.hg.1 | ENST00000534540.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009680.hg.1 | ENST00000534540.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009680.hg.1 | NR_038262.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009680.hg.1 | NR_038262.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009680.hg.1 | NR_038262.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009680.hg.1 | NR_038262.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009680.hg.1 | NR_038262.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009680.hg.1 | NR_038262.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009680.hg.1 | NR_038262.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015608.hg.1 | ENST00000362168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015608.hg.1 | ENST00000525865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015608.hg.1 | NR_029623.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015608.hg.1 | TC11001239.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1100009680.hg.1 | ENST00000362168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009680.hg.1 | ENST00000500447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009680.hg.1 | ENST00000500447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009680.hg.1 | ENST00000500447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009680.hg.1 | ENST00000500447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009680.hg.1 | ENST00000500447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009680.hg.1 | ENST00000528245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009680.hg.1 | ENST00000528245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009680.hg.1 | ENST00000528245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009680.hg.1 | ENST00000528245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009680.hg.1 | ENST00000528245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009680.hg.1 | ENST00000533920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009680.hg.1 | ENST00000533920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009680.hg.1 | ENST00000533920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009680.hg.1 | ENST00000533920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009680.hg.1 | ENST00000533920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009680.hg.1 | ENST00000534540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009680.hg.1 | ENST00000534540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009680.hg.1 | ENST00000534540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009680.hg.1 | NR_038262.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009680.hg.1 | NR_038262.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009680.hg.1 | NR_038262.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009680.hg.1 | NR_038262.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009680.hg.1 | NR_038262.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009680.hg.1 | NR_038262.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009680.hg.1 | NR_038262.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3310171 | NR_029623.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3310171 | NR_029623.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3310171 | NR_029623.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3310171 | NR_029623.1 | human_transcriptome |

MIR210
Genomic Map