Assay Details
| Gene Symbol: MIR99B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029843.1 | 1 | 38 | 64 | ||
| Gene Symbol: SPACA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017026299.1 | XP_016881788.1 | 1 | 829 | 64 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SPACA6P antisense RNA | SPACA6P-AS | LINC01129 | Anti-sense | Chr. 19 - 51685363 - 51693456 on NCBI Build GRCh38 | |
| Downstream | microRNA let-7e | MIRLET7E |
LET7E, MIRNLET7E, hsa-let-7e, let-7e |
105.0 | Sense | Chr. 19 - 51692786 - 51692864 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1900008662.hg.1
This assay targets the same location as HTA transcript cluster(s) TC19000784.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017026299.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Chr array_18 19 21 | A_17_P31632847 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632842 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632826 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993853 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632857 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632831 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993880 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993879 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632823 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632848 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632833 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993871 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993878 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993866 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993872 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993855 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993856 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993881 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993865 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993863 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993862 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993869 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993859 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993870 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993860 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632829 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632855 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993874 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993867 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993877 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17155569 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993861 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993858 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993864 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632840 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993882 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993885 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632838 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993857 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632825 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993883 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993884 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632864 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993873 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993876 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632841 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632835 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993854 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632843 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993875 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632824 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993868 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993886 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31632843 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993866 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993878 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993859 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993865 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993883 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993861 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31632864 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31632826 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993868 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993877 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993858 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31632825 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993873 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31632848 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993879 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31632842 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993871 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993863 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993886 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31632840 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993876 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993881 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31632824 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993875 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31632831 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31632847 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993857 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31632841 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31632855 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993867 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632835 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993868 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632829 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993884 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993871 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632824 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993853 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993865 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632838 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993866 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632857 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632864 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993886 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993863 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993874 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993855 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993881 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993882 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993877 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993883 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993885 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993857 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993861 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632833 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632826 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993875 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993879 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993878 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993872 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993860 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993864 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632843 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993862 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993858 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993856 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993870 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632847 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632823 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993859 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993880 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632840 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993869 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632842 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632831 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632841 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993867 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17155569 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993854 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632855 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632848 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993873 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10993876 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31632825 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31632841 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993873 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993875 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993878 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993876 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31632855 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993868 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31632843 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993861 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993859 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31632864 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31632848 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993858 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31632831 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31632842 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31632847 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993881 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993867 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993857 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993883 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31632825 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993863 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993877 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31632826 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993886 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31632840 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993865 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31632824 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993871 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993866 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10993879 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.1 ST Array | 8030842 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008662.hg.1 | ENST00000384819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008662.hg.1 | NR_029843.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19011038.hg.1 | ENST00000384819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19011038.hg.1 | NR_029843.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19011038.hg.1 | TC19000784.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011268.hg.1 | ENST00000602324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011268.hg.1 | ENST00000602324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011268.hg.1 | ENST00000602324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011268.hg.1 | NR_108100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011268.hg.1 | NR_108100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011268.hg.1 | NR_108100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011268.hg.1 | ENST00000602324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011268.hg.1 | ENST00000602324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011268.hg.1 | ENST00000602324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011268.hg.1 | NR_108100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011268.hg.1 | NR_108100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011268.hg.1 | NR_108100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008662.hg.1 | ENST00000384819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008662.hg.1 | NR_029843.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900167627.hg.1 | ENST00000384819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900167627.hg.1 | NR_029843.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900167627.hg.1 | TC1900008662.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900167627.hg.1 | NR_029843.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900167627.hg.1 | ENST00000384819.1 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3310166 | NR_029843.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3310166 | NR_029843.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3310166 | NR_029843.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3310166 | NR_029843.1 | human_transcriptome |

MIR99B
Genomic Map