| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22004812.hg.1 |
NR_037471.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22004812.hg.1 |
TC22000258.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007199.hg.1 |
ENST00000579518.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007199.hg.1 |
NR_037471.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200150612.hg.1 |
ENST00000579518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200150612.hg.1 |
NR_037471.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200150612.hg.1 |
TC2200007199.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200150612.hg.1 |
ENST00000579518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200150612.hg.1 |
NR_037471.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007199.hg.1 |
ENST00000579518.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007199.hg.1 |
NR_037471.1 |
Refseq |