Assay Details
| Gene Symbol: MIR711 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031756.1 | 1 | 40 | 67 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | urocortin 2 | UCN2 |
SRP, UCN-II, UCNI, UR, URP |
-15.1kb | Sense | Chr. 3 - 48561718 - 48563768 on NCBI Build GRCh38 |
| Downstream | ubiquinol-cytochrome c reductase core protein I | UQCRC1 |
D3S3191, QCR1, UQCR1 |
20.0kb | Sense | Chr. 3 - 48598999 - 48609665 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0300011028.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03001383.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223182 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223157 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223189 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223173 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223153 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223113 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22357591 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22357560 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223123 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223164 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22357602 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223147 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223125 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223195 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223191 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22357585 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223119 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223188 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223117 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22357618 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22357607 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223142 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223138 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223171 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223163 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223190 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22357569 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223161 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223127 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223140 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223175 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223197 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223134 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223115 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223118 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223178 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223169 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223160 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223183 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223176 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223155 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223128 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223145 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223193 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223149 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223167 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223116 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223122 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223152 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223131 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223172 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22357591 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22357607 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223122 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223149 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223160 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223172 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223182 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223131 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223190 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223167 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223161 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223176 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223125 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22357560 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223157 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223175 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223155 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223117 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223118 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223178 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223152 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223115 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223191 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223169 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223193 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223164 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223123 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223138 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223145 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223163 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223171 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223183 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223113 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22357618 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223195 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223128 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223173 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223119 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223142 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223189 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223116 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223147 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223153 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223140 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223197 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22357569 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22357602 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223188 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22357585 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223134 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223127 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.1 ST Array | 8086985 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024176.hg.1 | ENST00000390201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024176.hg.1 | NR_031756.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024176.hg.1 | TC03001383.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0300180895.hg.1 | ENST00000328333.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300180895.hg.1 | ENST00000390201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300180895.hg.1 | ENST00000487017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300180895.hg.1 | NM_000094.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300180895.hg.1 | NR_031756.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300180895.hg.1 | ENST00000328333.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300180895.hg.1 | ENST00000390201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300180895.hg.1 | ENST00000487017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300180895.hg.1 | NM_000094.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300180895.hg.1 | NR_031756.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300180895.hg.1 | TC0300011028.hg.1 | Clariom D TC |

MIR711
Genomic Map