Assay Details
| Gene Symbol: LOC100335030 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033267.1 | 1 | 3012 | 136 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DnaJ heat shock protein family (Hsp40) member C22 | DNAJC22 | wus | -35.5kb | Sense | Chr. 12 - 49346917 - 49353684 on NCBI Build GRCh38 |
| Downstream | potassium voltage-gated channel subfamily H member 3 | KCNH3 |
BEC1, ELK2, Kv12.2 |
146.7kb | Sense | Chr. 12 - 49538541 - 49558312 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000373.hg.1
Set Membership: |
|

LOC100335030
Genomic Map