Assay Details
| Gene Symbol: LINC00514 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033861.1 | 2 - 3 | 998 | 69 | ||
| GenBank mRNA | AK098017.1 | - | 2 - 3 | 998 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
protein kinase, membrane associated tyrosine/threonine 1 |
PKMYT1 |
MYT1, PPP1R126 |
-9.5kb | Anti-sense | Chr. 16 - 2972791 - 2980539 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101929613 | LOC101929613 | LA16c-380H5.3 | 7.9kb | Sense | Chr. 16 - 2998955 - 3000813 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000092.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000571152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000571152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000571152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000571152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000571152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000571152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000571152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000571152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000571152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000571152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000571152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001678.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001678.hg.1 | TC16000092.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001679.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001679.hg.1 | TC16000092.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001680.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001680.hg.1 | TC16000092.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000571152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000571152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000571152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000571152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000571152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000571152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000571152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000571152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000571152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000571152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000571152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011333.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600188925.hg.1 | ENST00000571152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188925.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188925.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188925.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188925.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188925.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600188925.hg.1 | TC1600011333.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600188925.hg.1 | ENST00000571152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188925.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188925.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188925.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188925.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188925.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600188926.hg.1 | ENST00000571152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188926.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188926.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188926.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188926.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188926.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600188926.hg.1 | TC1600011333.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600188926.hg.1 | ENST00000571152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188926.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188926.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188926.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188926.hg.1 | ENST00000573465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188926.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600188924.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188924.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188924.hg.1 | NR_033861.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600188924.hg.1 | TC1600011333.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600188924.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188924.hg.1 | ENST00000572266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600188924.hg.1 | NR_033861.1 | Refseq |

LINC00514
Genomic Map