Assay Details
| Gene Symbol: MIR3129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036076.1 | 1 | 32 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | collagen type V alpha 2 chain | COL5A2 | EDSC | Sense | Chr. 2 - 189031915 - 189225301 on NCBI Build GRCh38 | |
| Downstream | WD repeat domain 75 | WDR75 |
NET16, UTP17 |
308.3kb | Anti-sense | Chr. 2 - 189441433 - 189475562 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0200015205.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02002613.hg.1
Set Membership: |
|

MIR3129
Genomic Map