| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08008115.hg.1 |
NR_038201.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08008115.hg.1 |
NR_038202.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08008115.hg.1 |
NR_038203.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08008115.hg.1 |
NR_038204.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08008115.hg.1 |
NR_038205.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08008115.hg.1 |
NR_038206.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08008115.hg.1 |
NR_038207.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08008115.hg.1 |
NR_038208.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08008115.hg.1 |
NR_038209.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08008115.hg.1 |
TC08000584.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0800157060.hg.1 |
ENST00000517437.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157060.hg.1 |
NR_038201.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157060.hg.1 |
NR_038202.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157060.hg.1 |
NR_038203.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157060.hg.1 |
NR_038204.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157060.hg.1 |
NR_038205.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157060.hg.1 |
NR_038206.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157060.hg.1 |
NR_038207.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157060.hg.1 |
NR_038208.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157060.hg.1 |
NR_038209.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157060.hg.1 |
ENST00000517437.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800157060.hg.1 |
NR_038201.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157060.hg.1 |
NR_038202.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157060.hg.1 |
NR_038203.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157060.hg.1 |
NR_038204.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157060.hg.1 |
NR_038205.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157060.hg.1 |
NR_038206.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157060.hg.1 |
NR_038207.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157060.hg.1 |
NR_038208.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157060.hg.1 |
NR_038209.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800157060.hg.1 |
TC0800008284.hg.1 |
Clariom D TC |