Assay Details
| Gene Symbol: C8orf37-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038202.1 | 1 - 2 | 57 | 86 | ||
| NR_038207.1 | 57 | 86 | ||||
| GenBank mRNA | GU295153.1 | - | 1 - 2 | 57 | 86 | |
| GU295160.1 | - | 1 - 2 | 57 | 86 | ||
| GU295161.1 | - | 57 | 86 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1298 | LINC01298 | C8orf69 | -52.5kb | Anti-sense | Chr. 8 - 95207007 - 95216374 on NCBI Build GRCh38 |
| Downstream | serine/arginine-rich splicing factor 3 pseudogene | LOC100500773 | 514.2kb | Anti-sense | Chr. 8 - 95946984 - 95948348 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000584.hg.1
Additional Information:
This assay is in the 5'UTR region of the GU295153.1,GU295160.1,GU295161.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08008107.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008107.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008107.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008107.hg.1 | NR_038207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008107.hg.1 | TC08000584.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000517437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000517437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000517437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000517437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000517437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000517437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000517655.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000517655.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000517655.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000614093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000614093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038204.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038204.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038204.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038204.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038204.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038206.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038206.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038206.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038206.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038206.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008108.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008108.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008108.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008108.hg.1 | NR_038207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008108.hg.1 | TC08000584.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008102.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008102.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008102.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008102.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008102.hg.1 | NR_038204.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008102.hg.1 | NR_038205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008102.hg.1 | NR_038206.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008102.hg.1 | NR_038207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008102.hg.1 | NR_038208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008102.hg.1 | NR_038209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008102.hg.1 | TC08000584.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008286.hg.1 | ENST00000519366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008286.hg.1 | ENST00000519366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008286.hg.1 | ENST00000519366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008286.hg.1 | ENST00000519366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008286.hg.1 | ENST00000519366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008286.hg.1 | ENST00000519366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008286.hg.1 | ENST00000519366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008286.hg.1 | ENST00000519366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008286.hg.1 | ENST00000519366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008286.hg.1 | ENST00000519366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157043.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157043.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157043.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157043.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157043.hg.1 | NR_038204.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157043.hg.1 | NR_038205.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157043.hg.1 | NR_038206.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157043.hg.1 | NR_038207.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157043.hg.1 | NR_038208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157043.hg.1 | NR_038209.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157043.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157043.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157043.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157043.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157043.hg.1 | NR_038204.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157043.hg.1 | NR_038205.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157043.hg.1 | NR_038206.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157043.hg.1 | NR_038207.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157043.hg.1 | NR_038208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157043.hg.1 | NR_038209.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157043.hg.1 | TC0800008284.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800157049.hg.1 | ENST00000517437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157049.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157049.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157049.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157049.hg.1 | NR_038207.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157049.hg.1 | ENST00000517437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157049.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157049.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157049.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157049.hg.1 | NR_038207.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157049.hg.1 | TC0800008284.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000517437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000517437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000517437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000517437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000517437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000517437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000517655.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000517655.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000517655.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000614093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000614093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038204.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038204.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038204.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038204.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038204.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038206.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038206.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038206.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038206.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038206.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038209.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157048.hg.1 | ENST00000517437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157048.hg.1 | ENST00000517655.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157048.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157048.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157048.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157048.hg.1 | NR_038207.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157048.hg.1 | ENST00000517437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157048.hg.1 | ENST00000517655.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157048.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157048.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157048.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157048.hg.1 | NR_038207.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157048.hg.1 | TC0800008284.hg.1 | Clariom D TC |

C8orf37-AS1
Genomic Map