Assay Details
| Gene Symbol: C8orf37-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038208.1 | 1 - 2 | 127 | 118 | ||
| GenBank mRNA | GU295162.1 | - | 2 - 3 | 127 | 118 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 8 open reading frame 37 | C8orf37 |
BBS21, CORD16, RP64, smalltalk |
-30.8kb | Anti-sense | Chr. 8 - 95244913 - 95270602 on NCBI Build GRCh38 |
| Downstream | serine/arginine-rich splicing factor 3 pseudogene | LOC100500773 | 146.4kb | Anti-sense | Chr. 8 - 95946984 - 95948348 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000584.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000517437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000517437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000517437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000517437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000517437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000517437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000517655.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000517655.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000517655.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000614093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | ENST00000614093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038204.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038204.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038204.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038204.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038204.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038206.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038206.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038206.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038206.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038206.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038207.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008284.hg.1 | NR_038209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008104.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008104.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008104.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008104.hg.1 | NR_038208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008104.hg.1 | TC08000584.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000517437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000517437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000517437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000517437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000517437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000517437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000517655.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000517655.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000517655.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000614093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | ENST00000614093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038202.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038204.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038204.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038204.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038204.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038204.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038206.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038206.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038206.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038206.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038206.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038207.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008284.hg.1 | NR_038209.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157044.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157044.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157044.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157044.hg.1 | NR_038208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157044.hg.1 | ENST00000521905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157044.hg.1 | NR_038201.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157044.hg.1 | NR_038203.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157044.hg.1 | NR_038208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157044.hg.1 | TC0800008284.hg.1 | Clariom D TC |

C8orf37-AS1
Genomic Map