Assay Details
| Gene Symbol: RAB5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252039.1 | NP_001238968.1 | 6 - 7 | 992 | 89 | |
| NM_004583.3 | NP_004574.2 | 5 - 6 | 776 | 89 | ||
| NM_201434.2 | NP_958842.1 | 6 - 7 | 893 | 89 | ||
| XM_011525090.1 | XP_011523392.1 | 5 - 6 | 715 | 89 | ||
| XM_011525091.1 | XP_011523393.1 | 5 - 6 | 839 | 89 | ||
| GenBank mRNA | AF141304.1 | - | 4 - 5 | 605 | 89 | |
| AF498938.1 | - | 4 - 5 | 536 | 89 | ||
| AK310234.1 | - | 6 - 7 | 936 | 89 | ||
| AL553867.3 | - | 5 - 6 | 731 | 88 | ||
| BC106039.1 | - | 5 - 6 | 735 | 4938169 | 89 | |
| BC114439.1 | - | 5 - 6 | 731 | 40083572 | 89 | |
| BT019484.1 | - | 4 - 5 | 536 | 89 | ||
| CR541901.1 | - | 4 - 5 | 536 | 89 | ||
| U11293.1 | - | 4 - 5 | 611 | 89 | ||
| U18420.1 | - | 5 - 6 | 671 | 89 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

RAB5C
Genomic Map