Assay Details
| Gene Symbol: NOTCH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017617.4 | NP_060087.3 | 24 - 25 | 4015 | 113 | |
| XM_011518717.2 | XP_011517019.2 | 21 - 22 | 3307 | 113 | ||
| GenBank mRNA | AB209873.1 | - | 16 - 17 | 2552 | 113 | |
| AF308602.1 | - | 24 - 25 | 4018 | 113 | ||
| M73980.1 | - | 24 - 25 | 4018 | 113 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001734.hg.1
Set Membership: |
|

NOTCH1
Genomic Map