| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001416.hg.1 |
NR_037177.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001416.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001416.hg.1 |
TC6_mann_hap4000080.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7001630.hg.1 |
NR_037177.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7001630.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7001630.hg.1 |
TC6_ssto_hap7000082.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001807.hg.1 |
NR_037177.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001807.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001807.hg.1 |
TC6_cox_hap2000096.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7001628.hg.1 |
NR_037177.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7001628.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7001628.hg.1 |
TC6_ssto_hap7000082.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001809.hg.1 |
NR_037177.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001809.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001809.hg.1 |
TC6_cox_hap2000096.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000696.hg.1 |
NR_037177.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000696.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000696.hg.1 |
TC6_apd_hap1000049.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001818.hg.1 |
NR_037177.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001818.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001818.hg.1 |
TC6_dbb_hap3000089.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001595.hg.1 |
NR_037177.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001595.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001595.hg.1 |
TC6_mcf_hap5000081.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001597.hg.1 |
NR_037177.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001597.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001597.hg.1 |
TC6_mcf_hap5000081.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000694.hg.1 |
NR_037177.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000694.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000694.hg.1 |
TC6_apd_hap1000049.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001770.hg.1 |
NR_037177.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001770.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001770.hg.1 |
TC6_qbl_hap6000087.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001772.hg.1 |
NR_037177.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001772.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001772.hg.1 |
TC6_qbl_hap6000087.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06004723.hg.1 |
NR_037177.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06004723.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06004723.hg.1 |
TC06000405.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06004724.hg.1 |
ENST00000499196.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06004724.hg.1 |
NR_037177.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06004724.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06004724.hg.1 |
TC06000405.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001820.hg.1 |
NR_037177.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001820.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001820.hg.1 |
TC6_dbb_hap3000089.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001418.hg.1 |
NR_037177.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001418.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001418.hg.1 |
TC6_mann_hap4000080.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06004726.hg.1 |
ENST00000499196.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06004726.hg.1 |
NR_037177.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06004726.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06004726.hg.1 |
TC06000405.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600152963.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600152963.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600152963.hg.1 |
TC0600007671.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600152969.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600152969.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600152969.hg.1 |
TC0600007671.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600152972.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600152972.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600152972.hg.1 |
TC0600007671.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600152973.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600152973.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600152973.hg.1 |
TC0600007671.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600152964.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600152964.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600152964.hg.1 |
TC0600007671.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600152968.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600152968.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600152968.hg.1 |
TC0600007671.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600152966.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600152966.hg.1 |
NR_037178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600152966.hg.1 |
TC0600007671.hg.1 |
Clariom D TC |