| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05007717.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05007717.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05007717.hg.1 |
TC05000555.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05007718.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05007718.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05007718.hg.1 |
TC05000555.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05007722.hg.1 |
ENST00000504107.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05007722.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05007722.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05007722.hg.1 |
TC05000555.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05007725.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05007725.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05007725.hg.1 |
TC05000555.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008415.hg.1 |
ENST00000504107.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008415.hg.1 |
ENST00000504107.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008415.hg.1 |
ENST00000504107.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008415.hg.1 |
ENST00000504107.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008415.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008415.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008415.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008415.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008415.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008415.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008415.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008415.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008415.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008415.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008415.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008415.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008415.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008415.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008415.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008415.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008415.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008415.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008415.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157591.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157591.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157591.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157591.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157591.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157591.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157591.hg.1 |
TC0500008415.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500008415.hg.1 |
ENST00000504107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008415.hg.1 |
ENST00000504107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008415.hg.1 |
ENST00000504107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008415.hg.1 |
ENST00000504107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008415.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008415.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008415.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008415.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008415.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008415.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008415.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008415.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008415.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008415.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008415.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008415.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008415.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008415.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008415.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008415.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008415.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008415.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008415.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157600.hg.1 |
ENST00000504107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157600.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157600.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157600.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157600.hg.1 |
ENST00000504107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157600.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157600.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157600.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157600.hg.1 |
TC0500008415.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500157606.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157606.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157606.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157606.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157606.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157606.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157606.hg.1 |
TC0500008415.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500157592.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157592.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157592.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157592.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157592.hg.1 |
ENST00000514186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157592.hg.1 |
NR_046089.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157592.hg.1 |
TC0500008415.hg.1 |
Clariom D TC |