Assay Details
| Gene Symbol: LINC00992 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046089.1 | 4 - 5 | 453 | 77 | ||
| GenBank mRNA | BC045192.1 | - | 4 - 5 | 461 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102467223 | LOC102467223 | -783.9kb | Anti-sense | Chr. 5 - 116743302 - 116762209 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC102467224 | LOC102467224 | 174.6kb | Sense | Chr. 5 - 117730361 - 118266062 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000555.hg.1
Set Membership: |
|

LINC00992
Genomic Map