| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011612.hg.1 |
ENST00000431637.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011612.hg.1 |
ENST00000431637.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011612.hg.1 |
ENST00000431637.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011612.hg.1 |
ENST00000431637.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011612.hg.1 |
NR_048550.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011612.hg.1 |
NR_048550.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011612.hg.1 |
NR_048550.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011612.hg.1 |
NR_048550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011612.hg.1 |
ENST00000431637.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011612.hg.1 |
ENST00000431637.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011612.hg.1 |
ENST00000431637.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011612.hg.1 |
ENST00000431637.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011612.hg.1 |
NR_048550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011612.hg.1 |
NR_048550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011612.hg.1 |
NR_048550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011612.hg.1 |
NR_048550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100185510.hg.1 |
ENST00000431637.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100185510.hg.1 |
NR_048550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100185510.hg.1 |
ENST00000431637.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100185510.hg.1 |
NR_048550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100185510.hg.1 |
TC0100011612.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100185507.hg.1 |
ENST00000431637.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100185507.hg.1 |
NR_048550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100185507.hg.1 |
ENST00000431637.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100185507.hg.1 |
NR_048550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100185507.hg.1 |
TC0100011612.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100185509.hg.1 |
ENST00000431637.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100185509.hg.1 |
NR_048550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100185509.hg.1 |
ENST00000431637.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100185509.hg.1 |
NR_048550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100185509.hg.1 |
TC0100011612.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100185511.hg.1 |
ENST00000431637.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100185511.hg.1 |
NR_048550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100185511.hg.1 |
ENST00000431637.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100185511.hg.1 |
NR_048550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100185511.hg.1 |
TC0100011612.hg.1 |
Clariom D TC |