Assay Details
| Gene Symbol: CRLF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073118.1 | 1 - 2 | 245 | 77 | ||
| GenBank mRNA | AK296238.1 | - | 1 - 2 | 156 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SUZ12 polycomb repressive complex 2 subunit pseudogene 1 | SUZ12P1 | SUZ12P | -27.3kb | Anti-sense | Chr. 17 - 30709608 - 30770050 on NCBI Build GRCh38 |
| Downstream |
ATPase family, AAA domain containing 5 |
ATAD5 |
C17orf41, ELG1, FRAG1 |
6.7kb | Anti-sense | Chr. 17 - 30830932 - 30896424 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK296238.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
G1/S transition of mitotic cell cycle
negative regulation of cell growth
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of JAK-STAT cascade
positive regulation of cell cycle arrest
protein binding
G1/S transition of mitotic cell cycle
negative regulation of cell growth
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of JAK-STAT cascade
positive regulation of cell cycle arrest
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000583527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000583527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000583527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000583527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000583527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000583805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000583805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000585268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000585268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000585268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | ENST00000585268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000583527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000583527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000583527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000583527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000583527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000583805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000583805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000585268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000585268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000585268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | ENST00000585268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010293.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700180956.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180956.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180956.hg.1 | NM_015986.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700180956.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700180956.hg.1 | TC1700010293.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700180956.hg.1 | NR_073118.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700180956.hg.1 | ENST00000324238.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180956.hg.1 | ENST00000578692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180956.hg.1 | NM_015986.3 | Refseq |

CRLF3
Genomic Map