Assay Details
| Gene Symbol: ATF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073163.1 | 4 - 5 | 461 | 96 | ||
| GenBank mRNA | AK298853.1 | - | 4 - 5 | 407 | 96 | |
| X57197.1 | - | 3 - 4 | 303 | 96 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100652999 | LOC100652999 | -19.8kb | Anti-sense | Chr. 12 - 53513984 - 53517608 on NCBI Build GRCh38 | |
| Downstream |
ATP synthase, H+ transporting, mitochondrial Fo complex subunit C2 (subunit 9) |
ATP5G2 | ATP5A | 121.7kb | Sense | Chr. 12 - 53665160 - 53677546 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

ATF7
Genomic Map