Assay Details
| Gene Symbol: ALOX15P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045985.1 | 2 - 3 | 194 | 115 | ||
| GenBank mRNA | DY655533.1 | - | 2 - 3 | 194 | 115 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4520-1 | MIR4520-1 |
MIR4520A, hsa-mir-4520-1, mir-4520-1 |
-2.4kb | Anti-sense | Chr. 17 - 6655440 - 6655509 on NCBI Build GRCh38 |
| Downstream | solute carrier family 13 member 5 | SLC13A5 |
EIEE25, NACT, mIndy |
25.8kb | Anti-sense | Chr. 17 - 6684713 - 6713421 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000076.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17000926.hg.1 | NR_045985.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17000926.hg.1 | TC17000076.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17000928.hg.1 | NR_045985.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17000928.hg.1 | TC17000076.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000293761.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000293761.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000293761.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000293761.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000293761.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000293761.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000293761.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000293761.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000293761.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000293761.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000293761.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000293761.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000293761.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000293761.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000293761.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000293761.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000293761.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000293761.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000293761.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000570836.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000570836.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000570836.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000570836.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000570836.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000570836.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000570836.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000570836.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000570836.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000570836.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000570836.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000570836.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000570836.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000570836.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000570836.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000570836.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000570836.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000570836.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000570836.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000570836.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000572265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000572265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000572265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000572265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000572265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000573740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000573740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000573740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000573740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000573740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000573740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000574640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000574640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000574640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000574640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000574640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000574640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000574640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000574640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000574640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000574640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000574640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000574640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000574640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000574640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000574640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000574640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000574640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000574640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000574640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000576394.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000576394.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000576394.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000576394.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000576572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000576572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000576572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000576572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000576572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | ENST00000576572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | NM_001140.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | NM_001140.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | NM_001140.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | NM_001140.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | NM_001140.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | NM_001140.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | NM_001140.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | NM_001140.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | NM_001140.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | NM_001140.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | NM_001140.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | NM_001140.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | NM_001140.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | NM_001140.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | NM_001140.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | NM_001140.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | NM_001140.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | NM_001140.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009520.hg.1 | NM_001140.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006705.hg.1 | ENST00000621574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006705.hg.1 | ENST00000621574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006705.hg.1 | ENST00000621574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006705.hg.1 | ENST00000621574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006705.hg.1 | ENST00000621574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006705.hg.1 | ENST00000621574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006705.hg.1 | ENST00000621574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006705.hg.1 | ENST00000621574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006705.hg.1 | NR_045985.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006705.hg.1 | NR_045985.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006705.hg.1 | NR_045985.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006705.hg.1 | NR_045985.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006705.hg.1 | NR_045985.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17000927.hg.1 | NR_045985.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17000927.hg.1 | TC17000076.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700146216.hg.1 | ENST00000621574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700146216.hg.1 | NR_045985.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700146216.hg.1 | TC1700006705.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700146216.hg.1 | ENST00000621574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700146216.hg.1 | NR_045985.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700146213.hg.1 | ENST00000621574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700146213.hg.1 | NR_045985.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700146213.hg.1 | TC1700006705.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700146213.hg.1 | ENST00000621574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700146213.hg.1 | NR_045985.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006705.hg.1 | ENST00000621574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006705.hg.1 | ENST00000621574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006705.hg.1 | ENST00000621574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006705.hg.1 | ENST00000621574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006705.hg.1 | ENST00000621574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006705.hg.1 | ENST00000621574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006705.hg.1 | ENST00000621574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006705.hg.1 | ENST00000621574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006705.hg.1 | NR_045985.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006705.hg.1 | NR_045985.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006705.hg.1 | NR_045985.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006705.hg.1 | NR_045985.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006705.hg.1 | NR_045985.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700146214.hg.1 | ENST00000621574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700146214.hg.1 | NR_045985.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700146214.hg.1 | TC1700006705.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700146214.hg.1 | ENST00000621574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700146214.hg.1 | NR_045985.1 | Refseq |

ALOX15P1
Genomic Map