Assay Details
| Gene Symbol: SNORA27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002575.1 | 1 | 56 | 88 | ||
| GenBank mRNA | AJ609439.1 | - | 1 | 56 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
small nucleolar RNA, C/D box 102 |
SNORD102 |
RNU102, Z18 |
-266.0 | Sense | Chr. 13 - 27255064 - 27255135 on NCBI Build GRCh38 |
| Downstream | RAS like family 11 member A | RASL11A | 14.8kb | Sense | Chr. 13 - 27270327 - 27273690 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000073.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3800 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3799 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29184334 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08737924 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08737924 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29184334 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08737963 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08737952 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08737960 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08737958 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08737923 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08737954 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08737924 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08737956 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_002575_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7968235 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7968234 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR13000747.hg.1 | ENST00000384323.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13000747.hg.1 | ENST00000466550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13000747.hg.1 | NR_002575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13000747.hg.1 | TC13000073.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008856.hg.1 | ENST00000363365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000272274.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000272274.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000272274.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000272274.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000272274.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000272274.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000272274.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000311549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000311549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000311549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000311549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000311549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000311549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000311549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000311549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000319826.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000319826.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000319826.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000319826.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000319826.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000319826.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000319826.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000319826.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000319826.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000326092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000326092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000326092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000326092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000326092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000326092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000326092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000326092.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000384323.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000384769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000461690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000461690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000461690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000461690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000461690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000461690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000461690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000466550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000466550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000466550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000466550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000466550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000466550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000473558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000473558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000473558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000473558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000483765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000483765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000483765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000483765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000483765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000485756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000485756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000485756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000485756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000485756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000485756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000485756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000493317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000493317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000493317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000493317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000493317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | ENST00000493317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | NR_002574.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006675.hg.1 | NR_002575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009448.hg.1 | ENST00000362604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300145540.hg.1 | ENST00000384323.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300145540.hg.1 | ENST00000466550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300145540.hg.1 | NR_002575.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300145540.hg.1 | ENST00000384323.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300145540.hg.1 | ENST00000466550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300145540.hg.1 | NR_002575.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300145540.hg.1 | TC1300006675.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600009448.hg.1 | ENST00000362604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000272274.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000272274.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000272274.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000272274.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000272274.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000272274.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000272274.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000311549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000311549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000311549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000311549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000311549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000311549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000311549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000311549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000319826.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000319826.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000319826.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000319826.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000319826.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000319826.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000319826.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000319826.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000319826.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000326092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000326092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000326092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000326092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000326092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000326092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000326092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000326092.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000384323.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000384769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000461690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000461690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000461690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000461690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000461690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000461690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000461690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000466550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000466550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000466550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000466550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000466550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000466550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000473558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000473558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000473558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000473558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000483765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000483765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000483765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000483765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000483765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000485756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000485756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000485756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000485756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000485756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000485756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000485756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000493317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000493317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000493317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000493317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000493317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | ENST00000493317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | NR_002574.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006675.hg.1 | NR_002575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008856.hg.1 | ENST00000363365.1 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3248811 | NR_002575.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3248811 | AJ609439.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3248811 | NR_002575.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3248811 | AJ609439.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3248811 | NR_002575.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3248811 | AJ609439.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3248811 | NR_002575.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3248811 | AJ609439.1 | human_transcriptome |

SNORA27
Genomic Map