Assay Details
| Gene Symbol: CNBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127192.1 | NP_001120664.1 | 4 - 5 | 635 | 76 | |
| NM_001127193.1 | NP_001120665.1 | 4 - 5 | 632 | 76 | ||
| NM_001127194.1 | NP_001120666.1 | 4 - 5 | 614 | 76 | ||
| NM_001127195.1 | NP_001120667.1 | 4 - 5 | 611 | 76 | ||
| NM_001127196.1 | NP_001120668.1 | 4 - 5 | 608 | 76 | ||
| NM_003418.4 | NP_003409.1 | 4 - 5 | 629 | 76 | ||
| GenBank mRNA | AK054592.1 | - | 4 - 5 | 554 | 76 | |
| AK292119.1 | - | 4 - 5 | 578 | 76 | ||
| AK298154.1 | - | 4 - 5 | 521 | 76 | ||
| AK314380.1 | - | 4 - 5 | 554 | 76 | ||
| AY436367.1 | - | 1 - 2 | 54 | 76 | ||
| BC000288.2 | - | 4 - 5 | 571 | 2819226 | 76 | |
| BC014911.1 | - | 4 - 5 | 535 | 3906502 | 76 | |
| BC093058.1 | - | 4 - 5 | 548 | 30721134 | 76 | |
| BT019613.1 | - | 3 - 4 | 426 | 76 | ||
| DQ091187.1 | - | 4 - 5 | 564 | 76 | ||
| DQ092366.1 | - | 4 - 5 | 543 | 76 | ||
| DQ092367.1 | - | 4 - 5 | 546 | 76 | ||
| KU178397.1 | - | 3 - 4 | 402 | 76 | ||
| KU178398.1 | - | 3 - 4 | 427 | 76 | ||
| KU178399.1 | - | 3 - 4 | 423 | 76 | ||
| M28372.1 | - | 4 - 5 | 525 | 76 | ||
More Information
Set Membership: |
|

CNBP
Genomic Map