Assay Details
| Gene Symbol: CNEP1R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281789.1 | NP_001268718.1 | 3 - 4 | 284 | 81 | |
| NM_153261.5 | NP_694993.2 | 4 - 5 | 335 | 81 | ||
| GenBank mRNA | AK095420.1 | - | 3 - 4 | 257 | 81 | |
| AK301610.1 | - | 4 - 5 | 300 | 81 | ||
| AK307132.1 | - | 3 - 4 | 331 | 81 | ||
| BC036683.2 | - | 4 - 5 | 264 | 5266084 | 81 | |
| CR933600.1 | - | 4 - 5 | 280 | 81 | ||
| FJ516381.1 | - | 3 - 4 | 212 | 81 | ||
| HY011229.1 | - | 3 - 4 | 285 | 81 | ||
More Information
Set Membership: |
|

CNEP1R1
Genomic Map