Assay Details
| Gene Symbol: ISCA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194279.3 | NP_919255.2 | 3 - 4 | 345 | 110 | |
| GenBank mRNA | AK290728.1 | - | 3 - 4 | 318 | 110 | |
| BC032893.2 | - | 3 - 4 | 300 | 5259721 | 110 | |
| BX248252.1 | - | 3 - 4 | 303 | 110 | ||
More Information
Set Membership: |
|

ISCA2
Genomic Map