Assay Details
| Gene Symbol: CSRP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300965.1 | NP_001287894.1 | 3 - 4 | 362 | 95 | |
| NM_001321.2 | NP_001312.1 | 3 - 4 | 387 | 95 | ||
| GenBank mRNA | AY349166.1 | - | 3 - 4 | 342 | 95 | |
| BC000992.2 | - | 3 - 4 | 335 | 3448693 | 95 | |
| BT019913.1 | - | 2 - 3 | 283 | 95 | ||
| BX461295.2 | - | 3 - 4 | 409 | 95 | ||
| CN408330.1 | - | 3 - 4 | 333 | 95 | ||
| U46006.1 | - | 2 - 3 | 283 | 95 | ||
| U57646.1 | - | 3 - 4 | 339 | 95 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001755.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000311083.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000311083.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000311083.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000311083.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000311083.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000311083.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000311083.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000311083.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000311083.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000546966.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000546966.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000546966.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000546966.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000546966.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000546966.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000546966.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000547435.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000547435.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000547435.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000547435.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000547435.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000547435.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000547557.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000547557.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000547557.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000548783.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000548783.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000548783.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000548783.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000548783.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000548783.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000548783.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000548783.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000548783.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000548783.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000551725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000551725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000551725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000552330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000552330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000552330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000552330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000552330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000552330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000552330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | ENST00000552330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | NM_001300965.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | NM_001300965.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | NM_001300965.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | NM_001300965.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | NM_001300965.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | NM_001300965.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | NM_001300965.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | NM_001300965.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | NM_001321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | NM_001321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | NM_001321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | NM_001321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | NM_001321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | NM_001321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | NM_001321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | NM_001321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011327.hg.1 | NM_001321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12023260.hg.1 | ENST00000311083.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12023260.hg.1 | ENST00000546966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12023260.hg.1 | ENST00000547435.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12023260.hg.1 | ENST00000552330.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12023260.hg.1 | NM_001321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12023260.hg.1 | TC12001755.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000311083.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000311083.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000311083.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000311083.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000311083.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000311083.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000311083.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000311083.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000311083.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000546966.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000546966.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000546966.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000546966.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000546966.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000546966.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000546966.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000547435.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000547435.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000547435.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000547435.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000547435.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000547435.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000547557.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000547557.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000547557.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000548783.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000548783.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000548783.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000548783.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000548783.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000548783.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000548783.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000548783.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000548783.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000548783.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000551725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000551725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000551725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000552330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000552330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000552330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000552330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000552330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000552330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000552330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | ENST00000552330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | NM_001300965.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | NM_001300965.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | NM_001300965.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | NM_001300965.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | NM_001300965.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | NM_001300965.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | NM_001300965.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | NM_001300965.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | NM_001321.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | NM_001321.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | NM_001321.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | NM_001321.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | NM_001321.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | NM_001321.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | NM_001321.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | NM_001321.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011327.hg.1 | NM_001321.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187864.hg.1 | ENST00000311083.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187864.hg.1 | ENST00000546966.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187864.hg.1 | ENST00000547435.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187864.hg.1 | ENST00000548783.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187864.hg.1 | ENST00000552330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187864.hg.1 | NM_001300965.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187864.hg.1 | NM_001321.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187864.hg.1 | ENST00000311083.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187864.hg.1 | ENST00000546966.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187864.hg.1 | ENST00000547435.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187864.hg.1 | ENST00000548783.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187864.hg.1 | ENST00000552330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187864.hg.1 | NM_001300965.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187864.hg.1 | NM_001321.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187864.hg.1 | TC1200011327.hg.1 | Clariom D TC |

CSRP2
Genomic Map