Assay Details
| Gene Symbol: CSRP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300965.1 | NP_001287894.1 | 4 - 5 | 495 | 68 | |
| NM_001321.2 | NP_001312.1 | 4 - 5 | 520 | 68 | ||
| GenBank mRNA | AB209321.1 | - | 1 - 2 | 4672 | 68 | |
| BC000992.2 | - | 4 - 5 | 468 | 3448693 | 68 | |
| BT019913.1 | - | 3 - 4 | 416 | 68 | ||
| CN408330.1 | - | 4 - 5 | 466 | 68 | ||
| U46006.1 | - | 3 - 4 | 416 | 68 | ||
| U57646.1 | - | 4 - 5 | 472 | 68 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

CSRP2
Genomic Map