Assay Details
| Gene Symbol: HAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297436.1 | NP_001284365.1 | 2 - 3 | 753 | 76 | |
| NM_001523.3 | NP_001514.2 | 2 - 3 | 756 | 76 | ||
| XM_011526884.2 | XP_011525186.1 | 2 - 3 | 740 | 76 | ||
| GenBank mRNA | AY916554.1 | - | 1 | 53 | 91 | |
| AY916555.1 | - | 1 - 2 | 53 | 76 | ||
| AY916556.1 | - | 1 - 2 | 53 | 76 | ||
| BC035837.1 | - | 2 - 3 | 5589083 | 76 | ||
| BX371636.2 | - | 3 - 4 | 536 | 76 | ||
| D84424.1 | - | 1 - 2 | 740 | 76 | ||
| U59269.1 | - | 2 - 3 | 732 | 76 | ||
More Information
Set Membership: |
|

HAS1
Genomic Map