Assay Details
| Gene Symbol: NINJ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001294345.1 | NP_001281274.1 | 2 - 3 | 297 | 99 | |
| NM_001294346.1 | NP_001281275.1 | 2 - 3 | 404 | 99 | ||
| NM_016533.5 | NP_057617.2 | 2 - 3 | 643 | 99 | ||
| GenBank mRNA | AF205633.1 | - | 2 - 3 | 318 | 99 | |
| BC057766.1 | - | 2 - 3 | 311 | 99 | ||
| BI597990.1 | - | 2 - 3 | 496 | 99 | ||
| BM128205.1 | - | 2 - 3 | 265 | 99 | ||
| CB995632.1 | - | 2 - 3 | 324 | 99 | ||
| CD695373.1 | - | 2 - 3 | 391 | 99 | ||
More Information
Set Membership: |
|

NINJ2
Genomic Map