Assay Details
| Gene Symbol: ETNK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297760.1 | NP_001284689.1 | 4 - 5 | 1115 | 88 | |
| NM_001297761.1 | NP_001284690.1 | 3 - 4 | 855 | 88 | ||
| NM_001297762.1 | NP_001284691.1 | 3 - 4 | 992 | 88 | ||
| NM_018208.3 | NP_060678.2 | 4 - 5 | 1115 | 88 | ||
| XM_011509714.2 | XP_011508016.1 | 3 - 4 | 355 | 88 | ||
| XM_011509715.1 | XP_011508017.1 | 3 - 4 | 660 | 88 | ||
| XM_017001632.1 | XP_016857121.1 | 3 - 4 | 443 | 88 | ||
| GenBank mRNA | AB073608.1 | - | 4 - 5 | 715 | 88 | |
| AK001623.1 | - | 4 - 5 | 827 | 88 | ||
| AK293426.1 | - | 3 - 4 | 563 | 88 | ||
| AK302145.1 | - | 3 - 4 | 592 | 88 | ||
| AK316493.1 | - | 3 - 4 | 351 | 88 | ||
| BC010082.2 | - | 4 - 5 | 936 | 4329734 | 88 | |
More Information
Set Membership: |
|

ETNK2
Genomic Map