| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008988.hg.1 |
ENST00000582578.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008988.hg.1 |
ENST00000582578.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008988.hg.1 |
ENST00000582578.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008988.hg.1 |
ENST00000582578.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008988.hg.1 |
ENST00000582578.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008988.hg.1 |
NM_001278515.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008988.hg.1 |
NM_001278515.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008988.hg.1 |
NM_001278515.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008988.hg.1 |
NM_001278515.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008988.hg.1 |
NM_001278515.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008988.hg.1 |
NM_001278515.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800161683.hg.1 |
ENST00000582578.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800161683.hg.1 |
NM_001278515.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800161683.hg.1 |
TC1800008988.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800161683.hg.1 |
ENST00000582578.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800161683.hg.1 |
NM_001278515.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008988.hg.1 |
ENST00000582578.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008988.hg.1 |
ENST00000582578.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008988.hg.1 |
ENST00000582578.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008988.hg.1 |
ENST00000582578.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008988.hg.1 |
ENST00000582578.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008988.hg.1 |
NM_001278515.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008988.hg.1 |
NM_001278515.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008988.hg.1 |
NM_001278515.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008988.hg.1 |
NM_001278515.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008988.hg.1 |
NM_001278515.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008988.hg.1 |
NM_001278515.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800161684.hg.1 |
ENST00000582578.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800161684.hg.1 |
NM_001278515.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800161684.hg.1 |
TC1800008988.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800161684.hg.1 |
ENST00000582578.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800161684.hg.1 |
NM_001278515.1 |
Refseq |