Assay Details
| Gene Symbol: TAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001292034.2 | NP_001278963.1 | 4 - 5 | 1947 | 101 | |
| NM_001292035.2 | NP_001278964.1 | 4 - 5 | 1982 | 101 | ||
| NM_015093.5 | NP_055908.1 | 6 - 7 | 2190 | 101 | ||
| XM_011535633.2 | XP_011533935.1 | 5 - 6 | 2629 | 101 | ||
| XM_017010591.1 | XP_016866080.1 | 5 - 6 | 2160 | 101 | ||
| XM_017010592.1 | XP_016866081.1 | 5 - 6 | 2697 | 101 | ||
| GenBank mRNA | AB018276.2 | - | 4 - 5 | 1944 | 101 | |
| AF241230.1 | - | 3 - 4 | 1849 | 101 | ||
| AK025538.1 | - | 2 - 3 | 310 | 101 | ||
| AK295750.1 | - | 3 - 4 | 1803 | 101 | ||
| AL117407.1 | - | 5 - 6 | 2089 | 101 | ||
| BC035910.2 | - | 4 - 5 | 1905 | 5492359 | 101 | |
More Information
Set Membership: |
|

TAB2
Genomic Map